Bel-Art™ Sterileware™ Pharma Scoop
Bel-Art™ Sterileware™ Pharma Scoop is ideal for sampling, production work and laboratory use and used by pharmaceutical, food, cosmetic, and chemical companies.
$715.41 - $1435.17
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
03411784
|
Bel-Art™
F369090030 |
30 mL |
Pack of 100 Each for $942.93
|
|
|||||
03411785
|
Bel-Art™
F369090060 |
60 mL |
Pack of 100 Each for $951.28
|
|
|||||
03411786
|
Bel-Art™
F369090125 |
125 mL |
Pack of 100 Each for $1,277.97
|
|
|||||
03411787
|
Bel-Art™
F369090250 |
250 mL |
Pack of 100 Each for $1,435.17
|
|
|||||
03411788
|
Bel-Art™
F369090500 |
500 mL |
Pack of 50 Each for $1,365.83
|
|
|||||
03411789
|
Bel-Art™
F369091000 |
1000 mL |
Pack of 25 Each for $802.52
|
|
|||||
03411790
|
Bel-Art™
F369092500 |
2500 mL |
Pack of 15 Each for $715.41
|
|
|||||
Description
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High impact polystyrene (HIPS) scoops heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing
- Shipped with a Certificate of Processing for Sterilization
Specifications
High-impact Polystyrene | |
Sterile |