missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Stiripentol | |
77°C | |
C14H18O3 | |
1 g | |
IBLNKMRFIPWSOY-FNORWQNLSA-N | |
(E)-1-(1,3-benzodioxol-5-yl)-4,4-dimethylpent-1-en-3-ol | |
5311454 | |
≥98.0% (GC) |
49763-96-4 | |
White | |
MFCD00869310 | |
4,4-Dimethyl-1-(3,4-methylenedioxyphenyl)-1-penten-3-ol, 1-(1,3-Benzodioxol-5-yl)-4,4-dimethyl-1-penten-3-ol | |
CC(C)(C)C(C=CC1=CC2=C(C=C1)OCO2)O | |
234.295 | |
234.30 | |
Crystalline Powder |
Chemical Identifiers
49763-96-4 | |
234.295 | |
IBLNKMRFIPWSOY-FNORWQNLSA-N | |
5311454 | |
CC(C)(C)C(C=CC1=CC2=C(C=C1)OCO2)O |
C14H18O3 | |
MFCD00869310 | |
4,4-Dimethyl-1-(3,4-methylenedioxyphenyl)-1-penten-3-ol, 1-(1,3-Benzodioxol-5-yl)-4,4-dimethyl-1-penten-3-ol | |
(E)-1-(1,3-benzodioxol-5-yl)-4,4-dimethylpent-1-en-3-ol |
Safety and Handling
RTECSNumber : SB3393000
TSCA : No
Recommended Storage : Freezer