Accuform™ STOPOUT™ 120/240 Circuit Breaker Lockout, Double Pole
Lockout devices control energy sources and ensure machine and equipment remain inoperable to prevent unexpected start-up or release of stored energy in accordance with OSHA 29 CFR 1910.147 requirements
Supplier:  Accuform™ KDD161
Catalog No. 19111147
Explore available promotions
Description
Description
  • Features a bar clamp design, allowing device to slide onto a breaker switch and lock securely into place
  • Lift handle open, slide onto breaker bar, then close handle down and secure with a padlock (sold separately)
  • Fast secure ratchet application and quick-release tab for removal
  • Metal jaws for optimum grip
  • Octagon peg allows specially designed tab tags to be secured directly to the device (sold separately)
  • Accommodates padlock shackles up to 9/32 in. diameter (sold separately)
Specifications
Specifications
Lockout Device
Red
Plastic