Accuform™ STOPOUT™ 120/240 Circuit Breaker Lockout, Single Pole
Lockout devices control energy sources and ensure machine and equipment remain inoperable to prevent unexpected start-up or release of stored energy in accordance with OSHA 29 CFR 1910.147 requirements
Supplier: Accuform™ KDD160
Description
- Features a bar clamp design, allowing device to slide onto a breaker switch and lock securely into place
- Lift handle open, slide onto breaker bar, then close handle down and secure with a padlock (sold separately)
- Fast secure ratchet application and quick-release tab for removal
- Metal jaws for optimum grip
- Octagon peg allows specially designed tab tags to be secured directly to the device (sold separately)
- Accommodates padlock shackles up to 9/32 in. diameter (sold separately)
Specifications
Lockout Device | |
Red | |
Plastic |