Accuform™ STOPOUT™ Ball Valve Lockout
Lockout devices control energy sources and ensure machine and equipment remain inoperable to prevent unexpected start-up or release of stored energy in accordance with OSHA 29 CFR 1910.147 requirements
$34.69 - $57.17
Description
- Works with ball valve handles either perpendicular or parallel to pipe and secures with padlock (sold separately)
- Includes label printed with OSHA Danger header and message that reads LOCKED OUT with space to write more information as needed
- Red octagon shape reinforces lockout status and conveys STOP
Specifications
Ball Valve Lockout | |
Plastic |