Accuform™ STOPOUT™ Gate Valve Lockout
Lockout devices control energy sources and ensure machine and equipment remain inoperable to prevent unexpected start-up or release of stored energy in accordance with OSHA 29 CFR 1910.147 requirements
Supplier: Accuform™ KDD471
Description
- Hinged clamshell housing is designed to fully enclose the valve handle to prevent unauthorized access
- Tough thermoplastic, polypropylene device is a bright hazard red color, and features a durable, impact-resistant aluminum hinge with permanent binding pin
- Includes DANGER LOCKED OUT labels (English, Spanish, and French) with space to write more information as needed
- Red octagon shape reinforces lockout status and conveys STOP
Specifications
Gate Valve Lockout | |
Plastic | |
2-1/2 to 5 in. |