Eppendorf™ Storage Boxes
Allows selection of the optimal box combinations for individual storage demands and provides safe storage for your sample tubes in a space-saving design.
$57.67 - $130.50
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Holds | Price | Quantity | |||||
13-683-530
|
Eppendorf™
0030140508 |
100 Cryogenic Tubes with Internal Threads |
Each for $95.24
|
|
|||||
13-683-532
|
Eppendorf™
0030140540 |
81 Cryogenic Tubes (3 mL) with Screw Cap |
Each for $65.26
|
|
|||||
13-683-533
|
Eppendorf™
0030140567 |
81 Cryogenic Tubes (4 to 5 mL) with Screw Cap |
Each for $65.26
|
|
|||||
13-683-535
|
Eppendorf™
0030140532 |
25 Tubes (5 mL) |
Each for $130.50
|
|
|||||
13-683-534
|
Eppendorf™
0030140524 |
64 Tubes (1 to 2 mL) |
Each for $95.24
|
|
|||||
13-683-536
|
Eppendorf™
0030140613 |
25 Tubes (5 mL) with Screw Cap |
Each for $65.26
|
|
|||||
13-683-537
|
Eppendorf™
0030140583 |
25 Tubes (15 mL) |
Each for $57.67
|
|
|||||
Description
- Flexible and optimized combination options for the different box formats
- Retains shape at freezing temperature of -86°C for sample storage or autoclaving for 20 minutes at 121°C for sterilization
- Reliable individual alpha-numeric laser labeled code assigned to each sample location
- Minimizes allocation errors
- Provides high-contrast readability
- Remains permanently
- Five additional inscription labels on lid enable user-specific labeling
- Fits all typical tube formats: cryogenic tubes, microcentrifuge tubes, 15 and 50 mL conical tubes and other laboratory vessels
- 15 mL and 50 mL conical tube boxes with ventilation slots for an evenly distributed freezing process
- Prevents tube damage or breakage, which is frequently observed when freezing long tube formats in closed boxes
- Compatible with common freezer rack systems and the different inner grid variants
Specifications
Storage Box | |
Polypropylene | |
Yes |