Epredia™ STP 120 Spin Tissue Processor
Process biological specimens from chemical fixation to paraffin infiltration with the Epredia™ STP 120 Spin Tissue Processor.
$63106.28 - $71939.66
Specifications
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
36-101-3476
|
Epredia™
813150 |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
36-101-3477
|
Epredia™
813160 |
N/A
|
N/A | ||||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Immediate and delayed start processing modes
- Programmable spinning speed of either 60 or 70 rpm; spinning can also be programmed off
- Programmable immersion time in each station (from 1 minute to 99 hours: 59 minutes)
- Basket capacity of 120 or 240 cassettes
- Battery backup system in case of power failure
Performance:
Physical Specifications:
Includes:
Specifications
0.0 |