missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Streptavidin eFluor™ 710 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

$149.00 - $312.00

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 710
Form Liquid
Product Line eBioscience™
Product Type Streptavidin Conjugate (Fluorescent)
View More Specs

Alert:

Contains Azide


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016423
View Documents
Invitrogen™
49431780
25 μg
Each for $149.00
 
5016424
View Documents
Invitrogen™
49431782
100 μg
Each for $312.00
 
Description

Description

Specifications

Specifications

0.2 mg/mL
Liquid
Streptavidin Conjugate (Fluorescent)
eFluor 710
eFluor 710
eBioscience™
Biotin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only