Learn More
Streptavidin, Immobilized on Agarose CL-4B, For affinity chromatography, MilliporeSigma™ Supelco™
streptavidin attached to 4% beaded cross-linked agarose via a C6-spacer (1.2 mg streptavidin/ml packed gel)
Supplier:  Merck Emd Millipore 858811ML
 View more versions of this product
Catalog No. 111004376
Only null left
Explore available promotions
Description
Description
Isolation of cell surface glycoprotein by biotinylated concanavalin A and immobilized streptavidin.
Specifications
Specifications
Affinity Chromatography
1 mL
Product Suggestions
Product Suggestions