missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ eBioscience™ Streptavidin PerCP-eFluor™ 710 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry. Streptavidin binds to biotin with high affinity.

$222.00 - $421.00

Specifications

Concentration 0.2 mg/mL
Conjugate PerCP-eFluor 710
Form Liquid
Product Line eBioscience™
Product Type Streptavidin Conjugate (Fluorescent)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016042
View Documents
Invitrogen™
46431780
25 μg
Each for $222.00
 
5016043
View Documents
Invitrogen™
46431782
100 μg
Each for $421.00
 
Description

Description

Specifications

Specifications

0.2 mg/mL
Liquid
Streptavidin Conjugate (Fluorescent)
PerCP-eFluor 710
PerCP-eFluor 710
eBioscience™
Biotin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only