Streptavidin Peroxidase Polymer, Ultrasensitive , MilliporeSigma™ Supelco™
Activated streptavidin (SA) and horseradish peroxidase (HRP) are covalently conjugated to a polymer backbone. Multiple active biomolecules on each polymer chain increase the biotin binding capacity and amplify the peroxidase enzyme signal.
Supplier:  Merck Emd Millipore S2438250UG
Catalog No. 111014655
Explore available promotions
Description
Description
Specifications
Specifications
250 μg
Buffered aq. soln.