Learn More
Streptomycin sulfate, Cell Culture Reagent, Thermo Scientific Chemicals
Streptomycin sulfate, CAS # 3810-74-0, is the sulfate salt form of streptomycin, and an aminoglycoside with antibacterial activity. It also shows a protein synthesis inhibitory activity.
Supplier: Thermo Scientific Chemicals J6129922
Description
Inhibits initiation and causes misreading of rRNA in protein synthesis Streptomycin sulfate is used to study the mechanisms of streptomycin resistance. It is used to inhibit bacterial contamination in cell culture applications in association with penicillin. It is an antibiotic drug initially used for tuberculosis treatment. It inhibits protein synthesis by binding to 30S ribosomal subunit, which induces the misreading of the genetic code. It is involved in the initiation of translation of DNA, thereby results in death for a susceptible bacterium. Further, it controls bacterial and fungal diseases of certain fruit, vegetables, seed, ornamental crops and algae in ornamental ponds and aquaria
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Streptomycin Sulfate | |
Powder | |
C42H84N14O36S3 | |
3894995 | |
Soluble in water. | |
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O | |
1457.376 | |
728.69 | |
White |
Streptomycin sulfate | |
3810-74-0 | |
MFCD00037023 | |
14,8826 | |
QTENRWWVYAAPBI-FFCQDDOVSA-N | |
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-( | |
124080941 | |
Cell Culture | |
100 g |
Chemical Identifiers
3810-74-0 | |
1457.376 | |
QTENRWWVYAAPBI-FFCQDDOVSA-N | |
2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-( |
C42H84N14O36S3 | |
MFCD00037023 | |
124080941 | |
CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O |
Safety and Handling
GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c
H302-H317-H335-H361
EINECSNumber : 223-286-0
RTECSNumber : WK4990000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only