missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Strontium acetate hemihydrate, 98%, Thermo Scientific Chemicals
$71.40 - $964.62
Chemical Identifiers
CAS | 14692-29-6 |
---|---|
Molecular Formula | Sr(OOCCH3)2·0.5H2O |
MDL Number | MFCD00198064 |
Synonym | Acetic acid strontium salt |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1614922
|
Thermo Scientific Chemicals
A1614922 |
100 g |
Each for $71.40
|
|
|||||
AAA1614936
|
Thermo Scientific Chemicals
A1614936 |
500 g |
Each for $238.62
|
|
|||||
AAA161490E
|
Thermo Scientific Chemicals
A161490E |
2500 g |
Each for $964.62
|
|
|||||
Description
PST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPST precursor solutions were prepared from lead (II) acetate,titanium tetraisopropoxide, and strontium acetate hemihydrate with solvents of acetic acid and 2-methoxyethanol.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
14692-29-6 | |
Sr(OOCCH3)2·0.5H2O | |
MFCD00198064 | |
Acetic acid strontium salt |
Specifications
14692-29-6 | |
Sr(OOCCH3)2·0.5H2O | |
100 g | |
Acetic acid strontium salt | |
214.72 (205.71 Anhydrous) | |
Strontium acetate hemihydrate |
150°C (in -0.5H2O) | |
MFCD00198064 | |
14,8836 | |
Soluble in water. | |
98% |
Safety and Handling
EINECSNumber : 208-854-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only