Learn More
Strontium chloride hexahydrate, 98%, Thermo Scientific Chemicals
$112.66 - $362.70
Chemical Identifiers
CAS | 10025-70-4 |
---|---|
Molecular Formula | Cl2H12O6Sr |
Molecular Weight (g/mol) | 266.61 |
MDL Number | MFCD00149865 |
InChI Key | AMGRXJSJSONEEG-UHFFFAOYSA-L |
IUPAC Name | strontium(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1676430
|
Thermo Scientific Chemicals
A1676430 |
250 g |
Each for $112.66
|
|
|||||
AAA167640B
|
Thermo Scientific Chemicals
A167640B |
1000 g |
Each for $362.70
|
|
|||||
Description
Strontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsStrontium chloride hexahydrate is a precursor to prepare strontium chromate which is used as a corrosion inhibitor for aluminum. It is involved in the synthesis of cadmium sulfide core photocatalytic nanoparticles. It is used as a red coloring agent in pyrotechnics. Furthermore, it is utilized in glass-making and metallurgy. In the medical field, its radioactive isotope strontium-89 is used for the treatment of bone cancer. It acts as a catalyst in the conversion of ketones and aldehydes to the corresponding gem-dihydroperoxides using aqueous hydrogen peroxide.
Solubility
Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
10025-70-4 | |
266.61 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] |
Cl2H12O6Sr | |
MFCD00149865 | |
strontium(2+) hexahydrate dichloride |
Specifications
10025-70-4 | |
Cl2H12O6Sr | |
14,8840 | |
AMGRXJSJSONEEG-UHFFFAOYSA-L | |
strontium(2+) hexahydrate dichloride | |
266.62 (158.53 Anhydrous) | |
1.65 | |
1.93 |
250 g | |
MFCD00149865 | |
Soluble in water. Slightly soluble in ethanol and acetone. Insoluble in ammonia. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Sr++] | |
266.61 | |
≥99% | |
Hygroscopic | |
Strontium chloride hexahydrate |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
H318-H335
EINECSNumber : 233-971-6
RTECSNumber : WK8575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only