Learn More
Strontium granules, 20mm max, 99.0% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04292914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Golden | |
Distilled dendritic pieces | |
(metals basis) | |
MFCD00134060 | |
25 g | |
CIOAGBVUUVVLOB-UHFFFAOYSA-N | |
strontium | |
5359327 | |
99.8% | |
Air and moisture sensitive |
Strontium | |
7440-24-6 | |
Sr | |
UN3208 | |
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic | |
[Sr] | |
87.62 | |
CHEBI:33324 | |
Ampule |
Chemical Identifiers
7440-24-6 | |
87.62 | |
CIOAGBVUUVVLOB-UHFFFAOYSA-N | |
5359327 | |
strontium |
Sr | |
MFCD00134060 | |
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic | |
CHEBI:33324 | |
[Sr] |
Safety and Handling
GHS H Statement
H261
In contact with water releases flammable gas.
P223-P231+P232-P280-P335+P334-P370+P378q-P501c
H260
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
EINECSNumber : 231-133-4
RTECSNumber : WK7700000
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only