missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Strontium, distilled dendritic pieces, 99.95% (metals basis), Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 03862406

Numéro de catalogue. AA3862406


Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Spécifications

Spécifications

Strontium
7440-24-6
Sr
UN3208
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
[Sr]
87.62
CHEBI:33324
Ampule
Distilled dendritic pieces
(metals basis)
MFCD00134060
5 g
CIOAGBVUUVVLOB-UHFFFAOYSA-N
strontium
5359327
99.95%
Air and moisture sensitive

Identifiants chimiques

7440-24-6
87.62
CIOAGBVUUVVLOB-UHFFFAOYSA-N
5359327
strontium
Sr
MFCD00134060
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
CHEBI:33324
[Sr]
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H261
In contact with water releases flammable gas.

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H260

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

missing translation for 'einecsNumber' : 231-133-4

missing translation for 'rtecsNumber' : WK7700000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Argon

FDS
Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only