missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Strontium granules, 19mm (0.76in) & down, 99% (metals basis), Thermo Scientific Chemicals

Strontium granules, 99%, Sr, CAS Number-7440-24-6, atom, granules, 19mm 0.76in & down metals basis, estroncio, strontium, chunks, pieces, dendritic, pieces, yzs2rpe8le, strontium, elemental, strontium, distilled dendritic pieces, unii-yzs2rpe8le, 100g, (metals basis), CHEBI:33324, White to Yellow

Supplier:  Thermo Scientific Chemicals 03578922

 View more versions of this product

Catalog No. AA3578922


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon with thin oil layer
Specifications

Specifications

White to Yellow
7440-24-6
Sr
UN3208
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
[Sr]
87.62
CHEBI:33324
19mm (0.76 in.) & down
Strontium granules
(metals basis)
MFCD00134060
100 g
CIOAGBVUUVVLOB-UHFFFAOYSA-N
strontium
5359327
99%
Air and moisture sensitive

Chemical Identifiers

7440-24-6
87.62
CIOAGBVUUVVLOB-UHFFFAOYSA-N
5359327
strontium
Sr
MFCD00134060
strontium, elemental, unii-yzs2rpe8le, yzs2rpe8le, estroncio, granules, 19mm 0.76in & down metals basis, strontium, distilled dendritic pieces, atom, pieces, strontium, chunks, pieces, dendritic
CHEBI:33324
[Sr]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H261
In contact with water releases flammable gas.

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H260

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

EINECSNumber : 231-133-4

RTECSNumber : WK7700000

TSCA : Yes

storageNote1 :

  • Packaged under argon with thin oil layer

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only