missing translation for 'onlineSavingsMsg'
Learn More

Strontium Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical

$215.56 - $10173.79

Chemical Identifiers

CAS 10042-76-9
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614111
View Documents
Spectrum Chemical Mfg Cor
S168025GM
25 g Poly Bottle
Each for $215.56
Only null left
 
18614112
View Documents
Spectrum Chemical Mfg Cor
S1680125GM
125 g Poly Bottle
Each for $319.06
Only null left
 
18614113
View Documents
Spectrum Chemical Mfg Cor
S1680500GM
500 g Poly Bottle
Each for $710.38
Only null left
 
18614114
View Documents
Spectrum Chemical Mfg Cor
S168025KGPL
2.5 kg Poly Pail
Each for $2,031.97
Only null left
 
18614115
View Documents
Spectrum Chemical Mfg Cor
S168012KG
12 kg Poly Pail
Each for $6,574.00
Only null left
 
18614116
View Documents
Spectrum Chemical Mfg Cor
S168050KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10042-76-9

Specifications

10042-76-9
5.0 to 7.0
99%
0.001
1
25 g
ACS
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.