missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Strychnine, 98%, Thermo Scientific Chemicals

$165.26 - $592.81

Chemical Identifiers

CAS 57-24-9
Molecular Formula C21H22N2O2
Molecular Weight (g/mol) 334.42
MDL Number MFCD00005941
InChI Key QMGVPVSNSZLJIA-FVWCLLPLSA-N
Synonym strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
PubChem CID 441071
ChEBI CHEBI:28973
IUPAC Name (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
SMILES O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158950250
SDS View Documents
Thermo Scientific Chemicals
158950250
25 g Glass bottle
Each for $165.26
 
AC158951000
SDS View Documents
Thermo Scientific Chemicals
158951000
100 g Glass bottle
Each for $592.81
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57-24-9
334.42
QMGVPVSNSZLJIA-FVWCLLPLSA-N
441071
(4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
C21H22N2O2
MFCD00005941
strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
CHEBI:28973
O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61

Specifications

57-24-9
100.0
White
98%
C21H22N2O2
25 g
strychnine, strychnidin-10-one, --strychnine, strychnin, 3h strychnine, unii-h9y79vd43j, sanaseed, estricnina, strychinos, strychnos
QMGVPVSNSZLJIA-FVWCLLPLSA-N
(4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one
441071
334.41
Fine Crystalline Powder
97.5
284.0°C to 286.0°C
Authentic
Glass bottle
MFCD00005941
15, 8985
Solubility in water: slightly soluble. Other solubilities: soluble in chlororform and hot ethanol,insoluble in diethyl ether
O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1C1=CC=CC=C61
334.42
CHEBI:28973
98%
(-)-Strychnine, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-319-7

RTECSNumber : WL2275000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only