Learn More
Styrene-4-sulfonic acid sodium salt hydrate, tech. 90%, Thermo Scientific Chemicals
$59.89 - $112.85
Chemical Identifiers
CAS | 2695-37-6 |
---|---|
Molecular Formula | C8H7NaO3S |
Molecular Weight (g/mol) | 206.19 |
MDL Number | MFCD00013379 MFCD00084449 |
InChI Key | XFTALRAZSCGSKN-UHFFFAOYSA-M |
Synonym | sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt |
PubChem CID | 3571582 |
IUPAC Name | sodium;4-ethenylbenzenesulfonate |
SMILES | [Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1168506
|
Thermo Scientific Chemicals
L1168506 |
5 g |
N/A
|
|
||||||
AAL1168514
|
Thermo Scientific Chemicals
L1168514 |
25 g |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2695-37-6 | |
206.19 | |
XFTALRAZSCGSKN-UHFFFAOYSA-M | |
3571582 | |
[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1 |
C8H7NaO3S | |
MFCD00013379 MFCD00084449 | |
sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt | |
sodium;4-ethenylbenzenesulfonate |
Specifications
2695-37-6 | |
C8H7NaO3S | |
5 g | |
sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt | |
[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1 | |
206.19 | |
206.20 (Anhydrous) | |
Technical |
>320°C | |
MFCD00013379 MFCD00084449 | |
3575931 | |
XFTALRAZSCGSKN-UHFFFAOYSA-M | |
sodium;4-ethenylbenzenesulfonate | |
3571582 | |
90% | |
Styrene-4-sulfonic acid sodium salt hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-266-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only