missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Styrene-4-sulfonic acid sodium salt, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04169030

 View more versions of this product

Catalog No. AA4169030


Explore available promotions

Description

Description

Styrene-4-sulfonic acid sodium salt is grafted on poly(ethersulfone) membrane to determine the fractionation of proteins with similar molecular weights. It is used as sodium based ion exchange membranes for membrane capacitive deionization (MCDI) for synthesis. It is used in the study of copolymerization characteristics of sodium styrenesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Styrene-4-sulfonic acid sodium salt
White
MFCD00013379 MFCD00084449
3575931
Soluble in water.
[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1
206.19
206.19
2695-37-6
C8H7NaO3S
250 g
sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt
XFTALRAZSCGSKN-UHFFFAOYSA-M
sodium;4-ethenylbenzenesulfonate
3571582
Powder

Chemical Identifiers

2695-37-6
206.19
XFTALRAZSCGSKN-UHFFFAOYSA-M
3571582
[Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1
C8H7NaO3S
MFCD00013379 MFCD00084449
sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt
sodium;4-ethenylbenzenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-266-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only