missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Styrene, 99.5% stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

$63.55 - $304.97

Chemical Identifiers

CAS 100-42-5
Molecular Formula C8H8
Molecular Weight (g/mol) 104.15
MDL Number MFCD00008612,MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
SMILES C=CC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA16199AD
View Documents
Thermo Scientific Chemicals
016199AD
50 mL
Each for $63.55
 
AA16199AK
View Documents
Thermo Scientific Chemicals
016199AK
250 mL
Each for $118.70
 
AA16199K2
View Documents
Thermo Scientific Chemicals
016199K2
1 L
Each for $304.97
 
Description

Description

Styrene is used in the preparation of polystyrene, which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include in refrigerator liners, in medical devices and in small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN) which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-42-5
104.15
PPBRXRYQALVLMV-UHFFFAOYSA-N
7501
C=CC1=CC=CC=C1
C8H8
MFCD00008612,MFCD00084450
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
CHEBI:27452

Specifications

100-42-5
Colorless
neutral
31°C (88°F)
C8H8
MFCD00008612,MFCD00084450
UN2055
Air and light sensitive
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PPBRXRYQALVLMV-UHFFFAOYSA-N
styrene
7501
104.15
Technical
Styrene, Stabilized with 4-tert-butylcatechol
-31°C
0.906 g/mL
145°C to 146°C
Pungent
1.546
50 mL
1071236
14,8860
Miscible with alcohols,ethers and carbon disulfide. Immiscible with water.
C=CC1=CC=CC=C1
104.15
CHEBI:27452
99.5%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H304-H361-H226-H332-H315-H319-H335
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319-H332-H335-H336-H351-H361d-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: STYRENE MONOMER, STABILIZED

EINECSNumber : 202-851-5

RTECSNumber : WL3675000

TSCA : Yes

storageNote1 : 2°C to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only