Learn More
Succinic Acid 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158745000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Succinic acid | |
185.0°C to 190.0°C | |
235.0°C | |
C4H6O4 | |
MFCD00002789 | |
Solubility in water: 80g/L (20°C). Other solubilities: 1 kg/l boiling water, 1 g/18.5 mL alcohol - 1 g/6.3 mL methanol, 1 g/36 mL acetone - 1 g/20 mL glycerol, practically insoluble in petroleum ether, benzene,, carbon disulfide and carbon tetrachloride | |
OC(=O)CCC(O)=O | |
118.09 | |
CHEBI:15741 | |
99% | |
206°C |
99% | |
110-15-6 | |
99% | |
HO2CCH2CH2CO2H | |
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
butanedioic acid | |
1110 | |
118.09 | |
500 g | |
Plastic bottle |
Chemical Identifiers
110-15-6 | |
118.09 | |
KDYFGRWQOYBRFD-UHFFFAOYSA-N | |
1110 | |
butanedioic acid |
C4H6O4 | |
MFCD00002789 | |
succinic acid, amber acid, asuccin, dihydrofumaric acid, bernsteinsaure, katasuccin, wormwood acid, succinate, ethylenesuccinic acid, 1,2-ethanedicarboxylic acid | |
CHEBI:15741 | |
OC(=O)CCC(O)=O |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 203-740-4
RTECSNumber : WM4900000
TSCA : TSCA
RUO – Research Use Only