missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Succinic acid, disodium salt, 99%, anhydrous, Thermo Scientific Chemicals
$48.90 - $103.63
Chemical Identifiers
CAS | 150-90-3 |
---|---|
Molecular Formula | C4H4Na2O4 |
Molecular Weight (g/mol) | 162.052 |
MDL Number | MFCD00002790 |
InChI Key | ZDQYSKICYIVCPN-UHFFFAOYSA-L |
Synonym | disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 |
PubChem CID | 9020 |
ChEBI | CHEBI:63675 |
IUPAC Name | disodium;butanedioate |
SMILES | C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC208741000
|
Thermo Scientific Chemicals
208741000 |
100 g | Plastic bottle |
Each for $48.90
|
|
|||||
AC208745000
|
Thermo Scientific Chemicals
208745000 |
500 g | Plastic bottle |
Each for $103.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
150-90-3 | |
162.052 | |
ZDQYSKICYIVCPN-UHFFFAOYSA-L | |
9020 | |
disodium;butanedioate |
C4H4Na2O4 | |
MFCD00002790 | |
disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 | |
CHEBI:63675 | |
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] |
Specifications
150-90-3 | |
White | |
Authentic | |
Plastic bottle | |
NaO2CCH2CH2CO2Na | |
100 g | |
15, 8808 | |
Solubility in water: 300g/L (25°C). | |
C(CC(=O)[O-])C(=O)[O-].[Na+].[Na+] | |
162.052 | |
CHEBI:63675 | |
0.99 | |
Succinic acid, disodium salt, 0.99 |
>300.0°C | |
0.3% max. (120°C) | |
0.99 | |
C4H4Na2O4 | |
MFCD00002790 | |
02, 606 | |
disodium succinate, sodium succinate, disodium butanedioate, butanedioic acid, disodium salt, soduxin, succinic acid disodium salt, jantaran sodny czech, unii-v8zgc8isr3, succinic acid, disodium salt, fema no. 3277 | |
ZDQYSKICYIVCPN-UHFFFAOYSA-L | |
disodium;butanedioate | |
9020 | |
162.05 | |
Crystalline Powder |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 205-778-7
RTECSNumber : WM7751000
TSCA : TSCA
RUO – Research Use Only