missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinic Acid, FCC, 99-100.5%, Spectrum™ Chemical

S1686, 110-15-6, C4H6O4

$500.76 - $1655.26

Chemical Identifiers

CAS 110-15-6
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002789
InChI Key KDYFGRWQOYBRFD-UHFFFAOYSA-N
IUPAC Name butanedioic acid
SMILES OC(=O)CCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18607050
View Documents
Spectrum Chemical Mfg Cor
S1686500GM
500 g Poly Bottle
Each for $500.76
 
18607051
View Documents
Spectrum Chemical Mfg Cor
S168625KG
2.5 kg Poly Pail
Each for $1,655.26
 
Description

Description

Spectrum™ Chemical Succinic Acid, FCC is used as a flavoring agent for food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

110-15-6
118.09
KDYFGRWQOYBRFD-UHFFFAOYSA-N
OC(=O)CCC(O)=O
C4H6O4
MFCD00002789
butanedioic acid

Specifications

110-15-6
C4H6O4
KDYFGRWQOYBRFD-UHFFFAOYSA-N
butanedioic acid
99.0 to 100.5%
500 g
Poly Bottle
1
MFCD00002789
OC(=O)CCC(O)=O
118.09
FCC
0.00025
SDS
missing translation for 'documents'

missing translation for 'documents'