missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Succinimidyl 3-(2-Pyridyldithio)Propionate (SPDP)
The succinimidyl ester chemically reacts with primary amines and after mild reduction of the dithiol, it can react form a disulfide with other thiols.
Supplier: Invitrogen™ S1531
Description
Crosslinking reagents are useful for probing the spatial relationships and interactions within and between biomolecules.Specifications
Amine-Sulfhydryl | |
SPDP | |
No | |
Upon receipt store at -20°C protected from light. | |
Chemical Labeling | |
NHS Ester, Pyridyldisulfide | |
Solid | |
DMF, DMSO | |
No |
By Thiols | |
312.36 | |
6.8 AÌ | |
Yes | |
Heterobifunctional | |
Short (<10 AÌ) | |
100 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.