Learn More
Succinonitrile, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0503736
Description
Succinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSuccinonitrile dispersed with ionic liquids entrapped in a host polymer was used to constitute the gel polymer electrolytes. It was used as dopant to investigate the ionic conductivity and X-ray absorption spectroscopic results for lithium and copper salt doped succinonitrile.
Solubility
Slightly soluble in methanol and water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
Specifications
Succinonitrile | |
52°C to 56°C | |
132°C (269°F) | |
C4H4N2 | |
500 g | |
1098380 | |
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil | |
IAHFWCOBPZCAEA-UHFFFAOYSA-N | |
butanedinitrile | |
8062 | |
98% |
110-61-2 | |
265°C to 267°C | |
Odorless | |
MFCD00001949 | |
UN3439 | |
14,8872 | |
Slightly soluble in methanol and water. | |
N#CCCC#N | |
80.09 | |
80.09 |
Chemical Identifiers
110-61-2 | |
80.09 | |
IAHFWCOBPZCAEA-UHFFFAOYSA-N | |
8062 | |
N#CCCC#N |
C4H4N2 | |
MFCD00001949 | |
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil | |
butanedinitrile |
Safety and Handling
GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 203-783-9
RTECSNumber : WN3850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only