Learn More
Succinonitrile, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 171181000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Succinonitrile | |
110-61-2 | |
Colorless | |
132°C | |
99% min. (GC) | |
C4H4N2 | |
MFCD00001949 | |
02, 615 | |
15, 9003 | |
Solubility in water: 12.8%. Other solubilities: soluble in acetone,chloroform and dioxane,slightly soluble in ethanol,benzene,ether and,carbon disulfide | |
N#CCCC#N | |
80.09 | |
80.09 | |
Liquid |
99+% | |
57°C | |
265°C to 267°C | |
Authentic | |
Glass bottle | |
NCCH2CH2CN | |
100 g | |
04,241 | |
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil | |
IAHFWCOBPZCAEA-UHFFFAOYSA-N | |
butanedinitrile | |
8062 | |
99+% |
Chemical Identifiers
110-61-2 | |
80.09 | |
IAHFWCOBPZCAEA-UHFFFAOYSA-N | |
8062 | |
N#CCCC#N |
C4H4N2 | |
MFCD00001949 | |
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil | |
butanedinitrile |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 203-783-9
RTECSNumber : WN3850000
TSCA : TSCA
RUO – Research Use Only