missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinonitrile, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 171185000

 View more versions of this product

Catalog No. AC171185000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Succinonitrile
110-61-2
Colorless
132°C
99% min. (GC)
C4H4N2
MFCD00001949
02, 615
15, 9003
Solubility in water: 12.8%. Other solubilities: soluble in acetone,chloroform and dioxane,slightly soluble in ethanol,benzene,ether and,carbon disulfide
N#CCCC#N
80.09
80.09
Liquid
99+%
57°C
265°C to 267°C
Authentic
Glass bottle
NCCH2CH2CN
500 g
04,241
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
IAHFWCOBPZCAEA-UHFFFAOYSA-N
butanedinitrile
8062
99+%

Chemical Identifiers

110-61-2
80.09
IAHFWCOBPZCAEA-UHFFFAOYSA-N
8062
N#CCCC#N
C4H4N2
MFCD00001949
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
butanedinitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 203-783-9

RTECSNumber : WN3850000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only