missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Succinonitrile 99.0+%, TCI America™

$39.79 - $418.68

Chemical Identifiers

CAS 110-61-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00001949
InChI Key IAHFWCOBPZCAEA-UHFFFAOYSA-N
Synonym succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
PubChem CID 8062
IUPAC Name butanedinitrile
SMILES N#CCCC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S010925G
View Documents
TCI America
S010925G
25 g
Each for $39.79
 
S0109100G
View Documents
TCI America
S0109100G
100 g
Each for $103.81
 
S0109500G
View Documents
TCI America
S0109500G
500 g
Each for $418.68
 
Description

Description

Specifications

Chemical Identifiers

110-61-2
80.09
IAHFWCOBPZCAEA-UHFFFAOYSA-N
8062
N#CCCC#N
C4H4N2
MFCD00001949
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
butanedinitrile

Specifications

110-61-2
159°C
MFCD00001949
3439
IAHFWCOBPZCAEA-UHFFFAOYSA-N
butanedinitrile
8062
≥99.0% (GC)
Succinonitrile
White
C4H4N2
25 g
succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
N#CCCC#N
80.09
80.09
Crystals
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2771&(2)-1526

RTECSNumber : WN3850000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'