missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sucrose, (Crystalline), MP Biomedicals

This is a slightly less pure sucrose than our 190271. It does not reduce Fehling's solution, and is suitable for most routine applications

Supplier:  MP Biomedicals Inc 0290297825

 View more versions of this product

Catalog No. ICN90297825


Explore available promotions

Description

Description

  • Its a simple carbohydrate.
  • It is used in density gradients, Dielectrophoretic Field-Flow Fractionation running buffer.
  • It is a main component of media and animal diet.
  • Sucrose is a simple carbohydrate, a disaccharide composed of glucose and fructose.
  • It is hydrolyzed to glucose and fructose by dilute acids and by invertase, a yeast enzyme
Specifications

Specifications

Sucrose
C12H22O11
25 lb.
CZMRCDWAGMRECN-PWPRYFECNA-N
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
5988
342.3
57-50-1
MFCD00006626
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
342.30
CHEBI:17992
Crystalline

Chemical Identifiers

57-50-1
342.30
CZMRCDWAGMRECN-PWPRYFECNA-N
CHEBI:17992
C12H22O11
MFCD00006626
5988
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions