missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sucrose octaacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 419792500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sucrose octaacetate | |
>78.0°C | |
1.2800g/mL | |
0.1% max. | |
98% | |
C28H38O19 | |
1.28 | |
sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate | |
ZIJKGAXBCRWEOL-SAXBRCJISA-N | |
[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate | |
31340 | |
98% |
126-14-7 | |
Cream-White to White | |
250.0°C (1.0 mmHg) | |
Authentic | |
Plastic Bottle | |
250 g | |
15,9013 | |
Solubility in water: slightly soluble. Other solubilities: soluble in acetic acid,acetone,benzene,90g/l in ethanol | |
CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C | |
678.59 | |
678.59 | |
Powder |
Chemical Identifiers
126-14-7 | |
678.59 | |
sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate | |
[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate |
C28H38O19 | |
ZIJKGAXBCRWEOL-SAXBRCJISA-N | |
31340 | |
CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C |
Safety and Handling
EINECSNumber : 204-772-1
RUO – Research Use Only