missing translation for 'onlineSavingsMsg'
Learn More

Sudan IV, Spectrum™ Chemical

$214.43 - $3700.70

Chemical Identifiers

CAS 85-83-6
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614146
View Documents
Spectrum Chemical Mfg Cor
SU12025GM
25 g Amber Glass Bottle
Each for $214.43
Only null left
 
18614145
View Documents
Spectrum Chemical Mfg Cor
SU120100GM
100 g Amber Glass Bottle
Each for $532.43
Only null left
 
18614144
View Documents
Spectrum Chemical Mfg Cor
SU1201KG
1 kg Poly Pail
Each for $3,700.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

85-83-6

Specifications

85-83-6
Amber Glass Bottle
Reagent
1
25 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.