missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfadoxin 98.0+%, TCI America™

Supplier:  TCI America S089925G

 View more versions of this product

Catalog No. S089925G


Explore available promotions

Description

Description

Specifications

Specifications

Sulfadoxin
198°C
C12H14N4O4S
25 g
PJSFRIWCGOHTNF-UHFFFAOYSA-N
4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide
17134
310.33
Crystalline Powder
2447-57-6
White-Yellow
MFCD00792890
sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil
COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC
310.33
CHEBI:9329
≥98.0% (HPLC,T)

Chemical Identifiers

2447-57-6
310.33
PJSFRIWCGOHTNF-UHFFFAOYSA-N
17134
4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzene-1-sulfonamide
C12H14N4O4S
MFCD00792890
sulfadoxine, sulforthomidine, sulphadoxine, fanasil, sulphormethoxine, sulfadoxin, 4-amino-n-5,6-dimethoxypyrimidin-4-yl benzenesulfonamide, sulfadoxina, sulfadoxinum, fanzil
CHEBI:9329
COC1=NC=NC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DA9500000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions