missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfamethazine, 99%, Thermo Scientific Chemicals
$46.74 - $122.52
Chemical Identifiers
CAS | 57-68-1 |
---|---|
Molecular Formula | C12H14N4O2S |
Molecular Weight (g/mol) | 278.33 |
MDL Number | MFCD00006066 |
InChI Key | ASWVTGNCAZCNNR-UHFFFAOYSA-N |
Synonym | sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin |
PubChem CID | 5327 |
ChEBI | CHEBI:102265 |
IUPAC Name | 4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide |
SMILES | CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1927614
|
Thermo Scientific Chemicals
A1927614 |
25 g |
Each for $46.74
|
|
|||||
AAA1927622
|
Thermo Scientific Chemicals
A1927622 |
100 g |
Each for $122.52
|
|
|||||
Description
Sulfamethazine blocks the synthesis of dihydrofolic acid by inhibiting dihydropteroate synthase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSulfamethazine is an antibacterial. Sulfamethazine blocks the synthesis of dihydrofolic acid by inhibiting dihydropteroate synthase.Sulfamethazine is an antibiotic used to treat bronchitis, prostatitis and urinary tract infections. It is used in disposition and depletion kinetic studies. It is used to develop detection techniques for quantification in fluids such as cowsâ milk, honey and swine urine. Sulfamethazine is an antimicrobial sulfur drug that blocks the synthesis of dihydrofolic acid by inhibiting the enzyme dihydropteroate synthase. Sulfamethazine is a competitive inhibitor of bacterial para-aminobenzoic acid.
Solubility
Soluble in DMSO (56 mg/mL at 25°C), water (<1mg/mL at 25°C), and ethanol (<1mg/mL at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
57-68-1 | |
278.33 | |
ASWVTGNCAZCNNR-UHFFFAOYSA-N | |
5327 | |
4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide |
C12H14N4O2S | |
MFCD00006066 | |
sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin | |
CHEBI:102265 | |
CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C |
Specifications
57-68-1 | |
C12H14N4O2S | |
25 g | |
Soluble in DMSO (56mg/mL at 25°C),water (<1mg/mL at 25°C),and ethanol (<1mg/mL at 25°C). | |
CC1=CC(=NC(=N1)NS(=O)(=O)C2=CC=C(C=C2)N)C | |
278.33 | |
CHEBI:102265 | |
99% |
198°C to 201°C | |
MFCD00006066 | |
sulfamethazine, sulfadimidine, sulfadimethyldiazine, sulfamezathine, sulfadimerazine, sulphamethazine, sulphamezathine, sulfadimezine, sulphadimidine, sulfadimesin | |
ASWVTGNCAZCNNR-UHFFFAOYSA-N | |
4-amino-N-(4,6-dimethylpyrimidin-2-yl)benzenesulfonamide | |
5327 | |
278.34 | |
Sulfamethazine |
Safety and Handling
EINECSNumber : 200-346-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only