missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfamethoxazole 98.0+%, TCI America™

Supplier:  TCI America S036125G

Catalog No. S036125G


Explore available promotions

Description

Description

Specifications

Specifications

Sulfamethoxazole
170°C
C10H11N3O3S
25 g
JLKIGFTWXXRPMT-UHFFFAOYSA-N
4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
5329
253.28
Crystalline Powder
723-46-6
White
MFCD00010546
sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin
CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N
253.276
CHEBI:9332
≥98.0% (HPLC,T)

Chemical Identifiers

723-46-6
253.276
JLKIGFTWXXRPMT-UHFFFAOYSA-N
5329
4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
C10H11N3O3S
MFCD00010546
sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin
CHEBI:9332
CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N
Safety and Handling

Safety and Handling

RTECSNumber : WP0700000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions