missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfanilic Acid Anhydrous (Crystalline/Certified ACS), Fisher Chemical

Supplier:  Fisher Chemical FLA296500

Catalog No. A296-500



Explore available promotions

Description

Description

Specifications

Specifications

Sulfanilic Acid Anhydrous
287.8°C
1.49g/mL
98.0 to 102.0%
C6H7NO3S
MFCD00007886
sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
HVBSAKJJOYLTQU-UHFFFAOYSA-N
4-aminobenzenesulfonic acid
8479
173.19
Certified ACS
121-57-3
Blue
0.01% max.
Poly Bottle
4-NH2C6H4SO3H
500 g
0.02% max. Insoluble in Na2CO3
C1=CC(=CC=C1N)S(=O)(=O)O
173.186
CHEBI:27500
98.0 to 102.0%
Solid

Chemical Identifiers

121-57-3
173.186
HVBSAKJJOYLTQU-UHFFFAOYSA-N
8479
4-aminobenzenesulfonic acid
C6H7NO3S
MFCD00007886
sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
CHEBI:27500
C1=CC(=CC=C1N)S(=O)(=O)O
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions