missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfapyridine 98.0+%, TCI America™
Supplier: TCI America S007125G
Specifications
Sulfapyridine | |
190°C | |
C11H11N3O2S | |
25 g | |
GECHUMIMRBOMGK-UHFFFAOYSA-N | |
4-amino-N-pyridin-2-ylbenzenesulfonamide | |
5336 | |
249.29 | |
Crystalline Powder |
144-83-2 | |
White-Yellow | |
MFCD00038036 | |
sulfapyridine, sulphapyridine, 2-sulfapyridine, sulfidin, sulfidine, 2-sulfanilamidopyridine, streptosilpyridine, coccoclase, eubasinum, piridazol | |
C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N | |
249.288 | |
CHEBI:132842 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
144-83-2 | |
249.288 | |
GECHUMIMRBOMGK-UHFFFAOYSA-N | |
5336 | |
4-amino-N-pyridin-2-ylbenzenesulfonamide |
C11H11N3O2S | |
MFCD00038036 | |
sulfapyridine, sulphapyridine, 2-sulfapyridine, sulfidin, sulfidine, 2-sulfanilamidopyridine, streptosilpyridine, coccoclase, eubasinum, piridazol | |
CHEBI:132842 | |
C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N |
Safety and Handling
RTECSNumber : DA9625000
TSCA : Yes