CHEMetrics Sulfide Activator Solution
The Sulfide Activator Solution accessory pack (A-9500) supplies enough solution to perform at least 200 tests.
Supplier:  CHEMetrics A9500
Catalog No. 13-299-209C
Explore available promotions
Description
Description
Specifications
Specifications
Liquid
6
Sulfide Activator Solution
Sulfide CHEMets, Vacu-vials and VACUettes test kit
6 x 10 mL
Product Suggestions
Product Suggestions