missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfolane, 99%, Thermo Scientific Chemicals

Miscible with water, acetone, toluene

Supplier:  Thermo Scientific Chemicals A134660B

 View more versions of this product

Catalog No. AAA134660B


Explore available promotions

Description

Description

Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

Solubility
Miscible with alcohol, water, acetone, toluene and dilute mineral acids. Slightly miscible with octanes, olefins and naphthenes.

Notes
Miscible with water, acetone, tolueneIncompatible with strong oxidizing agents.
Specifications

Specifications

Tetramethylene Sulfone
25°C to 28°C
285°C
99%
1.4825
1000 g
Hygroscopic
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
HXJUTPCZVOIRIF-UHFFFAOYSA-N
thiolane 1,1-dioxide
31347
120.17
126-33-0
1.2606
166°C (330°F)
C4H8O2S
MFCD00005484
107765
14,8955
Miscible with alcohol,water,acetone,toluene and dilute mineral acids. Slightly miscible with octanes,olefins and naphthenes.
O=S1(=O)CCCC1
120.17
CHEBI:74794
99%

Chemical Identifiers

126-33-0
120.17
HXJUTPCZVOIRIF-UHFFFAOYSA-N
31347
O=S1(=O)CCCC1
C4H8O2S
MFCD00005484
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
CHEBI:74794
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 204-783-1

RTECSNumber : XN0700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only