missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfur powder, sublimed, -100 mesh, 99.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03339436

 View more versions of this product

Catalog No. AA3339436


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sulphur
Yellow
500 g
S
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
[S]
32.06
CHEBI:26833
Weak
7704-34-9
Sublimed Powder
99.50%
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
sulfur
5362487
248°C (478°F)
Pale yellow powder

Chemical Identifiers

7704-34-9
32.06
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
sulfur
S
MFCD00085316
polymeric, lac, ad usum externum, acmc-20a47y, rubbervulcanizationagentis ot
CHEBI:26833
[S]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319-H500

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-722-6

RTECSNumber : WS4250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only