missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfur trioxide trimethylamine complex, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 159351000

 View more versions of this product

Catalog No. AC159351000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Sulfur trioxide trimethylamine complex
232.0°C
Authentic
Glass bottle
(CH3)3N·SO3
100 g
sulfur trioxide trimethylamine complex, sulfur trioxide trimethylamine, sulfur trioxide-trimethylamine complex, sulphur trioxide trimethylamine 1:1, sulfur trioxide-trimethylamine, trimethylamine sulfur trioxide, sulfur trioxide; trimethylamine, 1n1&1&&so3 complex, trimethylamine, compd. with sulfur trioxide 1:1, trimethylamine, compound with sulphur trioxide
DXASQZJWWGZNSF-UHFFFAOYSA-N
N,N-dimethylmethanamine;sulfur trioxide
222852
95%
3162-58-1
White
95%
C3H9NO3S
MFCD00012421
01,1128; 08,466
Solubility in water: insoluble. Other solubilities: soluble in hot water, ethanol
CN(C)C.O=S(=O)=O
139.17
139.17
Powder

Chemical Identifiers

3162-58-1
139.17
DXASQZJWWGZNSF-UHFFFAOYSA-N
222852
CN(C)C.O=S(=O)=O
C3H9NO3S
MFCD00012421
sulfur trioxide trimethylamine complex, sulfur trioxide trimethylamine, sulfur trioxide-trimethylamine complex, sulphur trioxide trimethylamine 1:1, sulfur trioxide-trimethylamine, trimethylamine sulfur trioxide, sulfur trioxide; trimethylamine, 1n1&1&&so3 complex, trimethylamine, compd. with sulfur trioxide 1:1, trimethylamine, compound with sulphur trioxide
N,N-dimethylmethanamine;sulfur trioxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 221-614-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only