missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.200 N (N/5), Ricca Chemical

Supplier:  Ricca Chemical Company 826016

 View more versions of this product

Catalog No. 826016


Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
7664-93-9,7732-18-5
0.200Normal
Natural Poly Bottle
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Colorless
97.0,1.0
100.0,1.04
500 mL
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Videos
SDS
Documents

Documents

Promotions

Promotions