missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.200 N (N/5), Ricca Chemical

Supplier:  Ricca Chemical Company 826032

 View more versions of this product

Catalog No. 8260-32


Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
7664-93-9,7732-18-5
0.200Normal
Natural Poly Bottle
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Colorless
97.0,1.0
100.0,1.04
1 L
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Videos
SDS
Documents

Documents

Promotions

Promotions