missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid, 1.00 N, Ricca Chemical
Supplier: Ricca Chemical Company R830000055E
Specifications
Sulfuric Acid | |
7664-93-9,7732-18-5 | |
Poly Drum | |
1.000 ± 0.001 N at 20°C | |
H2O4S | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 |
1N, TP Acid Digest Reagent | |
Odorless | |
Aqueous Solution | |
55 gal. | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(=O)(=O)O | |
98.072 | |
CHEBI:26836 |