missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric acid, 1N (0.5M) standard solution, Thermo Scientific Chemicals

$76.07 - $111.48

Specifications

Chemical Name or Material Sulfuric acid
Color Colorless
Name Note 1N (0.5M) Standard Solution
CAS 7664-93-9,7732-18-5
Concentration 0.998 to 1.002N (20°C)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124240010
View Documents Promotion Details
Thermo Scientific Chemicals
124240010
1 L Plastic bottle
Each for $76.07
 
AC124240025
View Documents
Thermo Scientific Chemicals
124240025
2.5 L Plastic bottle
Each for $111.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications

Specifications

Sulfuric acid
1N (0.5M) Standard Solution
0.998 to 1.002N (20°C)
Plastic bottle
H2SO4
MFCD00064589
15, 9104
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.07
Colorless
7664-93-9,7732-18-5
1.05g/mL
H2O4S
Liquid
01,470; 05,633; 06,558; 07,347; 09,441
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Pure
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only