missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sulfuric acid, 20% fuming, 18-24% free SO{3}, Thermo Scientific Chemicals

$104.96 - $783.89

Chemical Identifiers

CAS 8014-95-7
Molecular Formula H2O7S2
Molecular Weight (g/mol) 178.13
MDL Number MFCD00137792
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
Synonym oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
PubChem CID 24681
IUPAC Name sulfonylideneoxidane; sulfuric acid
SMILES O=S(=O)=O.OS(O)(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4554322
View Documents Promotion Details
Thermo Scientific Chemicals
04554322
100 g
Each for $104.96
 
AA4554336
View Documents Promotion Details
Thermo Scientific Chemicals
04554336
500 g
Each for $220.66
 
AA4554357
View Documents Promotion Details
Thermo Scientific Chemicals
04554357
4 x 500 g
Each for $783.89
 
Description

Description

Sulfuric acid is a strong dehydrating agent and is applied in many applications like domestic acidic drain cleaners, electrolytes in lead-acid batteries and various cleansing agents. It is useful for mineral processing, fertilizer manufacture, petroleum purification, wastewater processing and chemical synthesis. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide shows a useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8014-95-7
178.13
HIFJUMGIHIZEPX-UHFFFAOYSA-N
24681
O=S(=O)=O.OS(O)(=O)=O
H2O7S2
MFCD00137792
oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
sulfonylideneoxidane; sulfuric acid

Specifications

8014-95-7
Hygroscopic
100 g
Liquid
oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
HIFJUMGIHIZEPX-UHFFFAOYSA-N
sulfonylideneoxidane; sulfuric acid
24681
1.9
20% fuming, 18-24% free SO3
H2O7S2
MFCD00137792
Miscible with water.
O=S(=O)=O.OS(O)(=O)=O
178.13
Sulfuric acid; 15% fuming
Safety and Handling

Safety and Handling

P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H290-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only