missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sulfuric Acid (70%/Textile), Fisher Chemical
Supplier: Thermo Fisher Scientific Formerly Fairlawn SA1704
Specifications
Sulfuric Acid | |
10°C | |
290°C to 338°C | |
Pass Test | |
<0.001 mmHg at 20°C | |
4 L | |
Liquid | |
1830 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
Textile |
70% | |
7664-93-9,7732-18-5 | |
1.6015g/cm³ | |
Poly Bottle | |
69.0 to 71.0 % | |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
69.0 to 71.0% |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
DOTInformation : Class 8: Corrosive