Learn More
Sulfuric Acid, 96% Solution in Water, Extra Pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 133610010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sulfuric acid | |
96% Solution in Water | |
7664-93-9 | |
1.8300g/mL | |
1.83 | |
1 L | |
H2O4S | |
H2SO4 | |
01,470; 05,633; 06,558; 07,347; 09,441 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08 | |
Extra Pure |
Colorless | |
10.0°C | |
290.0°C | |
Glass Bottle | |
96% | |
21 mPa.s (25°C) | |
Oily Liquid | |
MFCD00064589 | |
15, 9104 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat) | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
96% |
Chemical Identifiers
7664-93-9 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only